missing translation for 'onlineSavingsMsg'
Learn More

α-Amylase from Porcine pancreas, MP Biomedicals™

Product Code. 11480302 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480302 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11480302 Supplier MP Biomedicals Supplier No. 0219123925

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

α-Amylase from Porcine pancreas ≥700 u/mg protein, Opaque, White Suspension

  • It is used to hydrolyze α bonds of α-linked polysaccharides, such as starch and glycogen.
  • It has been used in various plant studies, such as metabolism studies in Arabidopsis.
  • It is isolated from porcine pancreas is a glycoprotein.
  • It is a single polypeptide chain of ∽475 residues containing two SH groups and four disulfide bridges and a tightly bound Ca2+ necessary for stability. Chloride ions are necessary for activity and stability.
TRUSTED_SUSTAINABILITY

Specifications

Color Opaque White
pH 5.5 to 8.0 (activity range), 7 (optimum)
For Use With (Application) Molecular biology
Content And Storage 4°C
Source Porcine pancreas
Quantity 25 mg
Product Type alpha-Amylase
Synonym β-N-acetylglucosaminidase porcine placenta
Form Suspension
compliance-icons compliance-icons
Product Identifier
  • alpha-Amylase
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: calcium chloride, alpha-amylase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.