missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Bovine Fibrinogen

Product Code. 11424246 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5g
100g
Unit Size:
100g
5g
This item is not returnable. View return policy

Product Code. 11424246

Brand: MP Biomedicals™ 08820224

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable as a substrate for cell culture and in clotting assays

Prepared by extensive washing of cryoprecipitate followed by a buffer extraction to remove unwanted proteins and precipitation with ammonium sulfate. Lyophilized powder. Clottable protein >90%.

Physical Characteristics

  • Appearance: Off-white powder
  • Clottable Protein (anhydrous): ≥90%
  • Moisture: ≤5%
  • Reported: Total protein, pH
Certifications: ISO, cGMP

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Mammalian Cell Culture
Name Bovine Fibrinogen 90% Clottable from Bovine Plasma
pH Range 6.4
Quantity 5g
Source Bovine Plasma
Storage Requirements <-10°C
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.