missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Bovine Transferrin, (HOLO)

Product Code. 11472632 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11472632

Brand: MP Biomedicals™ 0215233590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for regulating iron levels in cell culture

Transferrin is a 77kDa plasma glycoprotein with the capacity to bind ferric ions and as such is a source of bio-available iron. The product is frequently used as a nutrient in cell and microbial culture and as a media supplement in the production of bio-pharmaceutical products. Transferrin has also been shown to function as a factor in the growth stimulation of many cell types, such as granulocytes, macrophages and endothelial cells. Chromatographically purified.

Physical characteristics:

  • Appearance: Reddish-brown powder
  • Purity: ≥95% w/w
  • Iron: ≥1200 mg/g protein
  • Moisture: ≤5%
  • pH (3%): 6.0 to 8.0
  • Solubility: Dissolves in <20 min.
Microbial evlauation and control:
  • Bioburden: ≤100cfu/g
  • Endotoxin: ≤1 EU/mg
  • Mycoplasma: Not detected
  • Virus: Not detected
Certifications: ISO9000, cGMP, EDQM

TRUSTED_SUSTAINABILITY

Specifications

pH Range 6 to 8
Quantity 500 mg
Source Bovine Serum
Storage Requirements 4°C
Product Type Bovine Transferrin (HOLO)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.