missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 0.25% Trypsin-EDTA (1X)

Product Code. 11494805 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494805 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11494805 Supplier MP Biomedicals™ Supplier No. 091689649
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

0.25% Trypsin and 1mM EDTA in modified HBSS w/o Ca++ and Mg++, 100 mL

  • Trypsin-EDTA solutions are used to release adherent cells from culture surfaces.
  • Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues.
  • The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.
  • Ethylenediaminetetracetic acid (EDTA), a chelating agent, is added to trypsin solutions to enhance enzymatic activity by neutralizing calcium and magnesium ions that enhance cellto-cell adhesion and obscure the peptide bonds on which trypsin acts.

Specifications

Content And Storage -20°C
Concentration 1:250
Product Type Cell Culture Dissociation Reagent
Sterility Sterile
Without Additives No Calcium, No Magnesium
Formulation 0.25% (w/v) Solution, 0.25% Trypsin
pH 7 to 7.3
Quantity 100 mL
Source Porcine (for Trypsin)

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.