missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Coconut Oil, Hydrogenated

Product Code. 11415512 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
450 g
Unit Size:
1lb
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415512 450 g 1lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11415512 Supplier MP Biomedicals™ Supplier No. 0290140401
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Coconut Oil, Hydrogenated, White Semi-solid

Coconut oil is a common ingredient of animal diet.

Specifications

Product Type Animal Diet
Color White
Format Semisolid
For Use With (Application) Coconut Oil is a common ingredient of animal diet
Quantity 450 g
Storage Requirements 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.