missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Pancreatic Islet Isolation Grade. Suggested for pancreatic islet isolation. Prepared to contain lower tryptic activity levels. Activity: ≥160U per mg solid. Unit Definition: One unit liberates peptides from collagen equivalent in ninhydrin color to 1.0μmol of L-leucine in 5 hr. at pH 7.4 and 37°C in presence of calcium ions. Dialyzed, lyophilized.
Specifications
Specifications
| Concentration | >160 U/mg |
| Color | Brown |
| For Use With (Application) | For tissue dissociation combined with other Enzymes such as Elastase, Trypsin/Papain |
| Content And Storage | 4°C |
| Source | Clostridium histolyticum |
| Quantity | 500 mg |
| Product Type | Collagenase Type 4 |
| Inhibitors | Cysteine, EDTA or o-phenanthroline, alpha2-macroglobulin - a large plasma glycoprotein, alpha2-macroglobulin, alpha1-antitrypsin |
| Form | Lyophilized Powder |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?