missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CSM-LEU-URA , Powdered Media

Product Code. 11387909 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
This item is not returnable. View return policy

Product Code. 11387909

Brand: MP Biomedicals™ 114520222

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media

  • Add 0.67g of CSM-Leu-Ura to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined double drop-out medium

Applications:

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L
For Use With (Application) For growth of Saccharomyces cerevisiae
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.