Learn More
MP Biomedicals™ Diaphorase from Porcine Heart
Diaphorase is used in a study to assess the protein-protein interactions in assembly of lipoic acid on 2-oxoacid dehydrogenases of aerobic metabolism
Brand: MP Biomedicals™ 0215084405
Code : NEW
Additional Details : CAS Number : 9001-18-7 Weight : 0.01000kg
Description
- CAS Number: 9001-18-7
- EC Number: 232-587-6
- MDL Number: MFCD00130947
- Physical Appearance: Pale yellow, freeze dried powder
- Specific Activity: Approx. 10U/mg protein
- Format: Freeze-dried powder in presence of sucrose
- Soluble in distilled water or dilute buffer
Specifications
25 U/mg | |
Study the assess the Protein-Protein interactions in assembly of Lipoic Acid on the 2-oxoacid Dehydrogenases of aerobic metabolism | |
Procine heart | |
Diaphorase |
Pale Yellow | |
0°C | |
5 mg | |
Lyophilized Powder |
Safety and Handling
- Diaphorase
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Diaphorase