missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ DOB 2% Galactose, 1% Raffinose, Powdered Media

Product Code. 11337799 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
227 g
Unit Size:
227g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337799 227 g 227g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11337799 Supplier MP Biomedicals™ Supplier No. 114025912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be used to create SD or SDA by combining with the appropriate amino acid mixture

Formulation

  • Use 36.7g per liter
  • Contents/liter: 1.7g YNB, 5g Ammonium Sulfate, 20g Galactose, 10g Raffinose

Yeast cell culture, Gene expression

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation YNB 1.7g/L, Ammonium Sulfate 5g/L, Galactose 20g/L, Raffinose 10g/L
For Use With (Application) Yeast cell culture, gene expression, used to create SD or SDA
Quantity 227 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.