missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ DOBA Medium, Powdered Media
Shop All MP Biomedicals Products
Click to view available options
Quantity:
454 g
1000 g
Unit Size:
1kg
454g
Description
Formulation
- Use 43.7g powder liter
- Contents/liter: DOB, 17g Agar-Y
Applications:
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Specifications
| Certifications/Compliance | Molecular biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Grade | Molecular biology |
| Product Type | Yeast Media |
| Formulation | Nitrogen Sources : Ammonium Sulfate 5g/L, Carbon Source: Dextrose 20g/L, Vitamins : Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg/L, Trace Elements: Boric Acid 0.5mg/L, Copper Sulfate 0.04mg/L, Potassium Iodide 0.1mg/L, Ferric Chloride 0.2mg/L, Manganese Sulfate 0.4mg/L, Sodium Molybdate 0.2mg/L, Zinc Sulfate 0.4mg/L, Salts: Potassium Phosphate, Monobasic 1000mg/L, Magnesium Sulfate 500mg/L, Sodium Chloride 100mg/L, Calcium Chloride 100mg/L, Agar 17g/L |
| For Use With (Application) | For Yeast cell culture, gene expression, transcription regulation |
| Quantity | 454 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction