missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ TeenPrep™ Adapter for 12 x 15 mL Tube Holder on FastPrep-24

Product Code. 10429774 Shop All MP Biomedicals Products
Change view
Click to view available options
Holds:
12 x 15 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
10429774 12 x 15 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10429774 Supplier MP Biomedicals™ Supplier No. 116002526
  • ONLINE EXCLUSIVE PRICE
    1540.00 €
    1457.40 € / Pack of 1
    Save 82.60 €
    5% Off
View all promotions
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TeenPrep™ Adapter is a medium-size adapter that holds 12 x 15 mL Lysing Matrix tubes that allows frozen samples to be immediately processed with minimal hands-on manipulation.

Medium-size adapter that holds 12 x 15 mL Lysing Matrix tubes. It allows frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes. A wide range of disposable 15 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in seconds.

  • Application Notes: TeenPrep™ 12 X 15 mL Adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications.
  • Capacity: 12 tubes
  • Sample Volume: Up to 24 g

Specifications

Product Type Large Sample Volume Adapter
Sample Size 24 g
Content And Storage 15°C to 30°C
For Use With (Application) DNA and RNA isolation, Enzyme isolation and Protein production, natural products isolation, food preparation for quality analysis
Holds 12 x 15 mL
Weight (English) 1.115 lb.
Weight (Metric) 506 g

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.