missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Flaxseed Oil

Product Code. 11421532 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
450 g
Unit Size:
1lb
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421532 450 g 1lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11421532 Supplier MP Biomedicals™ Supplier No. 0296043201
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Supplement for Animal Diet also know as Linseed Oil

  • CAS Number 8001-26-1
  • Refractive Index: n20/D 1.4795 (lit.)

Specifications

Product Type Animal Diet
Format Liquid
Quantity 450 g
Storage Requirements 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.