missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Ready for direct use
- Removes primers, nucleotides, enzymes, mineral oil, salts, and other impurities
- Applications: DNA Clean-up
Specifications
Specifications
| Sample Type | Post amplification PCR reaction mixture (oil-free) |
| For Use With (Application) | Isolation of DNA from PCR reactions, removal of primers, Nucleotides, Enzymes, Mineral Oil, salts and other impurities from DNA samples |
| Includes | GeneClean™ Turbo™ Salt Solution, Wash Solution, Cartridges, Catch Tubes and Elution Solution |
| No. of Reactions | 50 Preps |
| Sample Volume | Up to 100 μL PCR Reaction Solution |
| Sample Source | Gels and Solutions |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?