missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Glucose Oxidase

Product Code. 11491092 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 kU
250 kU
50 kU
500 kU
Unit Size:
10ku
250ku
500ku
50ku
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491092 50 kU 50ku
11481092 10 kU 10ku
11401102 250 kU 250ku
11411102 500 kU 500ku
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11491092 Supplier MP Biomedicals™ Supplier No. 0219519650
  • ONLINE EXCLUSIVE PRICE
    179.00 €
    169.05 € / 50ku
    Save 9.95 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

From Aspergillus niger

Type X. Activity: >100U/mg. Unit Definition: one unit oxidizes 1.0μmol of β-D-glucose to D-gluconic acid and H2O2 per minute at 35°C, pH 5.1.

Specifications

Concentration >100 U/mg
Color Yellow
pH 4 to 7
For Use With (Application) Glucose Oxidase can be utilized in the enzymatic determination of D-glucose in solution
Content And Storage -20°C
Source Aspergillus niger
Quantity 50 kU
Product Type Glucose Oxidase
Molecular Weight (g/mol) ≈160000
Inhibitors Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, 50% D-arabinose
Form Lyophilized Powder
Show More Show Less
compliance-icons
Product Identifier
  • glucose oxidase
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: glucose oxidase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.