missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Serum, Goat, Heat Inactivated

Product Code. 11485055 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485055 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11485055 Supplier MP Biomedicals™ Supplier No. 092939249
  • ONLINE EXCLUSIVE PRICE
    45.40 €
    43.05 € / 100mL
    Save 2.35 €
    5% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Serum is used as a supplement to cell culture media.

Goat Serum is used as a supplement to cell culture media. Goat Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health.

Specifications

Certifications/Compliance U.S. regulations
Color Golden Brown
Content And Storage -20°C
Species Goat
Product Type Goat Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method 0.2 μm filtration
Heat Inactivated Yes

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.