missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ LB Agar Medium Powder

Product Code. 11313269 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5000 g
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313269 5000 g 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11313269 Supplier MP Biomedicals™ Supplier No. 113002242
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Dehydrated
Grade Molecular biology
Product Type LB Agar
Formulation Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L
For Use With (Application) For growth of Escherichia coli and certified for plasmid and other bacterial cultures in liquid medium, cloning, culture media, microbial media
Quantity 5000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.