missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lipase from Porcine Pancreas

Product Code. 11383999 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11383999

Brand: MP Biomedicals™ 0210081710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Catalyzes hydrolysis of esters in aqueous solutions and synthesis of esters in non-aqueous solutions

Lipases are a subclass of esterases. They also produce hydroxyl and carboxylic groups through the hydrolysis of ester linkages in poly(ethylene terephthalate).

  • Synonyms: Steapsin, Glycerol ester hydrolase, Tween hydrolase, Tributyrase, PPL, Triacylglycerol acylhydrolase, Triacylglycerol lipase
  • CAS Number: 9001-62-1
  • EC Number: 232-619-9
  • MDL Number: MFCD00131509
  • Format: Tan powder
  • Specific Activity: >12,000U/g solid
  • Extinction Coefficient: E1%280=13.3
  • Isoelectric point (pI): Lipase A=4.9 and Lipase B=5.0
  • Partly soluble in water and insoluble in alcohol

Used industrially for resolution of chiral compounds and transesterification production of biodiesel.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Chiral Compounds Resolution, Transesterification Production of Biodiesel
Name Lipase
Quantity 10 g
Source Porcine Pancreas
Storage Requirements 0°C
Form Powder
compliance-icons
Product Identifier
  • Lipase
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Lipase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.