missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix G
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 x 2 mL Tubes
50 x 2 mL Tubes
Unit Size:
Pack of 100
Pack of 50
Description
Silicon carbide has relatively low density and high strength and hardness, and therefore is effective at breaking tough, hard, or brittle cell membranes. Useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.
- Impact-resistant 2 mL tube contains 1.6 mm Silicon Carbide particles and 2 mm glass beads
- With brown caps
- Specific Gravity: Silicon Carbide: 3.20; Glass beads: 2.55
- Nucleic Acid Isolation and Purification
Specifications
Specifications
| Capacity | 2 mL |
| Sample Type | Spores, Animal Tissues, Fungi, Plants, Yeast |
| DNase | Dnase-free |
| For Use With (Application) | Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis |
| Matrix | Lysing matrix G |
| Molecular Formula | Silicon Carbide, SiC ; Glass, SiO2 |
| Quantity | 100 x 2 mL Tubes |
| RNase | RNase-free |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction