missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysozyme from Chicken Egg White, 20,000U/mg

Hydrolyzes b-1,4 glucosidic linkages between N-acetylmuramic acid and N-acetylglucosamine which occur in mucopeptide cell wall structure of certain microorganisms, such as Micrococcus lysodeikticus

Brand:  MP Biomedicals™ 0210083180


 View more versions of this product

Product Code. 11324029

  • 1635.00 € / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
  • CAS Number: 9001-63-2
  • EC Number: 232-620-4
  • MDL Number: MFCD00131557
  • Format: 3X crystallized, salt-Free, albumin-Free, lyophilized
  • Specific Activity: ≤20,000U/mg solid
  • Extinction Coefficient (E1%): 26.4 (280 nm)
  • Isoelectric point (pI): pH 11.0
  • pH: 3.6

Used for lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific antigen. Also acts as germinative agent of bacterial spores. In humans, lysozyme may mediate anti-tumor function of macrophages. Cartilage lysozyme has a role in cartilage calcification.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3X
11
0°C
100 g
14388
Crystallized, Lyophilized
White
Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen
Chicken egg white
Lysozyme
Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lysozyme

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Lysozyme

SDS
Documents

Documents

Certificates
Special Offers

Special Offers