missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
- CAS Number: 9001-63-2
- EC Number: 232-620-4
- MDL Number: MFCD00131557
- Format: 3X crystallized, salt-Free, albumin-Free, lyophilized
- Specific Activity: ≤20,000U/mg solid
- Extinction Coefficient (E1%): 26.4 (280 nm)
- Isoelectric point (pI): pH 11.0
- pH: 3.6
Used for lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific antigen. Also acts as germinative agent of bacterial spores. In humans, lysozyme may mediate anti-tumor function of macrophages. Cartilage lysozyme has a role in cartilage calcification.
Specifications
Specifications
| Concentration | 3X |
| Color | White |
| pH | 11 |
| For Use With (Application) | Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen |
| Content And Storage | 0°C |
| Source | Chicken egg white |
| Quantity | 5 g |
| Product Type | Lysozyme |
| Molecular Weight (g/mol) | 14388 |
| Inhibitors | Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes |
| Show More |
Safety and Handling
Product Identifier
- lysozyme
- Danger
- Respiratory sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: lysozyme
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?