missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ M9 Minimal Medium Salts (Maniatis Formulation), Powdered
Shop All MP Biomedicals ProductsDescription
- A commonly used base for preparing a defined synthetic medium for the growth of most E. coli host cells
- Powder is reconstituted in sterile purified water, (or water with melted purified agar added)
- Additional nutrients, such as glucose or other carbon source, casamino acids, vitamins, or antibiotics are added when the solution cools
- Add additional nutrients before plates are poured or media is inoculated with bacteria
Specifications
Specifications
| Certifications/Compliance | Molecular Biology Certified |
| Concentration | 56.4 g/L of a 5X Solution |
| Content And Storage | 15° to 30°C |
| Description | A 56.4 g sample of M9 Minimal Salts was dissolved in an aqueous solution to 1 liter, the pH adjusted to 7.4 (if needed), and the solution autoclaved to make a sterile 5X concentrate |
| Form | Powder |
| Media Type | M9 Minimal Medium |
| Product Type | Microbiology Media |
| Sterility | Sterile |
| Formulation | 33.9 g Na2HPO4; 15 g KH2PO4; 2.5 g NaCl; 5 g NH4Cl |
| For Use With (Application) | Preparing a Defined Synthetic Medium for The Growth of E. Coli Host |
| Show More |
Safety and Handling
Product Identifier
- M9 Minimal Medium Salts
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: ammonium chloride
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?