missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Newborn Bovine Serum
Used in cell culture to support the maturation of oocytes, to study the function of tubular epithelial cells, and to develop primary cultures of cells such as visceral adipocytes.
Brand: MP Biomedicals™ 092912154
Code : NEW
Additional Details : Weight : 0.01000kg
Specifications
Golden Brown | |
Newborn Calf Bovine Serum | |
0.2 μm filtration | |
30EU/mL | |
≤27mg/dL | |
No | |
7.1 to 8.2 |
-20°C | |
500 mL | |
Bovine Neonate | |
Cell culture and organ preservation applications | |
No | |
265 to 323mOsm/kg | |
4.4 to 7% |