missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Papain
Used in the enzymatic synthesis of amino acids, peptides, and other molecules
190.80 €
Specifications
Color | Off-White |
---|---|
pH | 6 to 7 |
For Use With (Application) | Enzymatic synthesis of Amino Acids, Peptides and other molecules |
Content And Storage | 4°C |
Source | Carica papaya latex |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11478180
|
MP Biomedicals™
0210256680 |
100 g |
190.80 €
100g |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sulfhydryl protease from Carica papaya latex. (A second protease, chymopapain, and a lysozyme have also been isolated from this same source.)
- Format: Powder
- Grade: Technical
- Molecular weight: 23,000
- Specific Activity:>1750 USP Units/mg
Specifications
Off-White | |
Enzymatic synthesis of Amino Acids, Peptides and other molecules | |
Carica papaya latex | |
23000 | |
Powder |
6 to 7 | |
4°C | |
Papain | |
AEBSF, Antipain, Cystatin, E-64, leupeptin, PMSF, TLCK and TPCK |