missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pepsin

Product Code. 11465322 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11465322

Brand: MP Biomedicals™ 0219536710

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

From porcine stomach mucosa

Purified by chromatography. Activity: 2000-2400U/mg protein. Unit Definition: One unit will produce a Δ A280 of 0.001 per minute at pH 2.0 and 37°C, measured as TCA-soluble products using hemoglobin as substrate. Lyophilized powder, essentially salt-free.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Used to digest Proteins and is commonly used in the preparation of Fab fragments from Antibodies
pH Range 1 to 3.5
Sample Type Porcine Stomach Mucosa
Content And Storage 0°C
Form Lyophilized
Quantity 10 g
Product Type Pepsin
compliance-icons compliance-icons
Product Identifier
  • Pepsin
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : pepsin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.