missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pepsin From Porcine Stomach Mucosa

Product Code. 11488220 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488220 500 g 500g
11468220 50 g 50g
11478220 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488220 Supplier MP Biomedicals™ Supplier No. 0210259890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pepsin cleaves peptides with an aromatic acid on either side of the peptide bond

Pepsin contains the “cathepsin” component which has milk curdling activity. It has a broad range of substrate activity and demonstrates an esterase acitivity.

  • CAS Number: 9001-75-6
  • EC Number: 232-629-3
  • MDL Number: MFCD00081840
  • Format: off white lyophilized powder
  • Specific activity: ≥1:10000

Used in preparation of Fab fragments from antibodies and to hydrolyze dry cervical samples in mice

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Molecular Biology, Proteomics
Molecular Weight (g/mol) ∽35000
Name Pepsin
pH Range 2.5 to 4.5
Quantity 500 g
Source Porcine
Storage Requirements 15° to 30°C
Form Powder
compliance-icons compliance-icons
Product Identifier
  • Pepsin, 1:10,000
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Pepsin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.