missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ PlantCon™ Plant Tissue Culture Container

Product Code. 11415005 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 Ea
10/Cs
200/Cs
Unit Size:
Pack of 10
Pack of 200
Product Code. Quantity unitSize
11415005 1 Ea Pack of 10
17345609 10/Cs Pack of 10
11435005 200/Cs Pack of 200
3 options
This item is not returnable. View return policy

Product Code. 11415005

Brand: MP Biomedicals™ 092672002

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Sterilized Base for PlantCon™ Plant Tissue Culture Container. Use PlantCon™ containers with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.

Clear polyvinyl chloride (PVC) dishes specifically designed for plant culture applications. Ideal for use in commercial horticulture or research settings. Allow exceptional light transmission and optimal growth. Reduce the time spent preparing an in vitro growth environment for plant cultures by eliminating the need for cleaning and sterilization.

Order Info

Base L x W x H: 3 3/4 x 3 3/4 x 1 3/8 in. (9.5 x 9.5 x 3.5cm). Covers and bases are sold separately. Sterile.

TRUSTED_SUSTAINABILITY

Specifications

Closure Type Covers Sold Separately (See 092672102)
Bottom Shape Square
Color Clear
Disposable Single-use
Material PVC
Shape Rectangular
Sterility Sterile
Type Cell Culture Dish
Quantity 1 Ea
Dimensions (L x W) 3.75 x 3.75 in. (9.5 x 9.5 cm)
Length (Metric) 9.5 mm
Width (Metric) 9.5 mm
Height (Metric) 7 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.