missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protein A from Staphylococcus aureus, 9–11mg

Product Code. 11466452 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
20 mg
Unit Size:
1mg
20mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466452 20 mg 20mg
11446452 1 mg 1mg
11456452 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466452 Supplier MP Biomedicals™ Supplier No. 0215005120
  • ONLINE EXCLUSIVE PRICE
    925.00 €
    876.75 € / 20mg
    Save 48.25 €
    5% Off
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

A highly stable cell surface receptor produced by several strains of Staphylococcus aureus which can conjugate with various reporter molecules without affecting the antibody binding site on the molecule

Consists of a single polypeptide chain of molecular weight 42 kDa, containing four repetitive domains rich in aspartic and glutamic acids but devoid of cysteine. Contains little or no carbohydrate and only 4 tyrosine residues and no tryptophans.

  • Gene Id: Staphylococcus aureus subsp. aureus NCTC 8325 …SAOUHSC_00069(3919448)
  • Binding Capacity: 9–11mg of human IgG per mg solid (Lit.)
  • Extinction Coefficient: E1%280 = 1.4 (Lit.)
  • Presentation: White frozen solid
  • Isoelectric Point: 4.85 to 5.10(Lit.)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein A from S. aureus
Quantity 20 mg
Form Lyophilized
For Use With (Application) ELISA, Immunohistochemistry, Western Blotting
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.