missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ QuickFlex™ Adapter for 96 x 2 mL Tube Holder on FastPrep 96

Product Code. 15232820 Shop All MP Biomedicals Products
Change view
Click to view available options
For Use With (Equipment):
FastPrep-96
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15232820 FastPrep-96 Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15232820 Supplier MP Biomedicals™ Supplier No. 116010570
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

QuickFlex™ adapter for 96 x 2 mL tube holder on FastPrep 96 is suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications.

  • Applications: QuickFlex™ 96 x 2 mL Adapter for FastPrep-96™ is a large sample volume adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • Product Description: High-throughput adapter for the FastPrep-96™ that holds 96 x 2 mL lysing matrix tubes. The ergonomic design of the adapter ensures an easy loading of lysing matrix tubes that remain securely sealed during the homogenization. It stand stable on the bench top and is commonly used as a tube rack. Ideally suited for high-throughput applications, up to 96 samples can be homogenized simultaneously. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • Capacity: 96 tubes
  • Sample Volume: Up to 20 g

Specifications

Product Type FastPrep-96 Adapters
Content And Storage Room Temperature
For Use With (Application) Extractions of nucleic acids, proteins, metabolites and other small molecules from samples
For Use With (Equipment) FastPrep-96

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.