missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Raffinose Powder

Product Code. 11347769 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
227 g
454 g
Unit Size:
227g
454g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347769 454 g 454g
11337769 227 g 227g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347769 Supplier MP Biomedicals™ Supplier No. 114010022
  • ONLINE EXCLUSIVE PRICE
    728.00 €
    689.85 € / 454g
    Save 38.15 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Pentahydrate powder is 98% pure

  • White powder

Yeast cell culture, Gene expression

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Additive D-(+)-Raffinose Pentahydrate
Form Powder
For Use With (Application) Yeast cell culture, gene expression
Quantity 454 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.