missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Raffinose Powder

Product Code. p-4474965 Shop All MP Biomedicals Products
Click to view available options
Quantity:
227 g
454 g
Unit Size:
227g
454g
This item is not returnable. View return policy

Product Code. 11347769

Brand: MP Biomedicals™ 114010022

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Pentahydrate powder is 98% pure

  • White powder

Yeast cell culture, Gene expression

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Additive D-(+)-Raffinose Pentahydrate
Form Powder
For Use With (Application) Yeast cell culture, gene expression
Quantity 454 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.