missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ RNaid™ Kit

Product Code. p-4696796 Shop All MP Biomedicals Products
Change view
Click to view available options
Product Type:
Slurry
Slurry plus SPIN Filter
Unit Size:
200 preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
11383109 Slurry 200 preps
11393129 Slurry plus SPIN Filter 200 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383109 Supplier MP Biomedicals™ Supplier No. 111007200
Add to basket
Add to basket
This item is not returnable. View return policy

RNaid™ Kit provides a simple and convenient way to isolate total RNA from agarose and polyacrylamide gels and from solutions.

  • Kit is designed to isolate and purify RNA (75 bases to 10 kb) from native and denaturing agarose and polyacrylamide gels.
  • Removal of unincorporated nucleotides, buffer salts, and other reagents from enzyme reactions.
  • The purified RNA is ready for reverse transcription and PCR, labeling and microarray analysis, and other common applications.
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Enzymatic Reactions, Solutions, Agarose Gels, Polyacrylamide Gels
For Use With (Application) To isolate, purify RNA (75 bases to 10 kb) from native, denaturing agarose/polyacrylamide gels
Product Type Slurry
Quantity 200 Preps
compliance-icons
Product Identifier
  • RNaid Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Diethyl Pyrocarbonate

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.