missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Roll & Grow™ Plating Beads

Product Code. 11462330 Shop All MP Biomedicals Products
Change view
Click to view available options
Holds:
2 Tubes
Unit Size:
Pack of 2
Product Code. Holds unitSize
11462330 2 Tubes Pack of 2
1 options
This item is not returnable. View return policy

Product Code. 11462330

Brand: MP Biomedicals™ 115000550

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Safely and easily spread yeast and bacterial cells

Roll & Grow™ Plating Beads are used to spread 25-400μL of bacterial or yeast cells across the surface of an agar plate, providing a safer, faster alternative to the traditional method of plating with a glass rod sterilized with ethanol and flame. Sterile Roll & Grow™ Plating Beads are provided in two skirted 50mL conical tubes. Between six and seven 4mm glass beads are used per plate for a total of approximately 160 standard 100mm plates.

  • Spread cells in just seconds per plate
  • Eliminate hazards of flame and ethanol
  • No need for fragile glass “hockey sticks”
  • Consistent, even colony distribution
  • Sterile and easy to use

Cell cycle; Cell proliferation; Cell metabolism; Cell viability; Cytotoxicity; Protein Trafficking; Bacteria cell culture; Cell preservation; Cell counting; Viability; Cryopreservation; Insect cell culture; Mammalian cell culture; Plant cell growth; Primary cell culture; Stem cell culture; Yeast cell culture

TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 Tubes
Material Glass
Product Type Roll and Grow Plating Beads
For Use With (Application) Cell cycle, cell proliferation, cell metabolism, cell viability, cytotoxicity, protein trafficking, bacteria cell culture, cell preservation, cell counting, viability, cryopreservation, insect cell culture, mammalian cell culture, plant cell growth
Includes Skirted 50mL conical tubes
Thickness 4mm
Storage Requirements 15° to 30°C
Sterility Sterile
Holds 2 Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.