missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Sodium Phosphate Buffer

Product Code. 12869211 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
60 mL
Unit Size:
60mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12869211 60 mL 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12869211 Supplier MP Biomedicals™ Supplier No. 116560205
  • ONLINE EXCLUSIVE PRICE
    52.05 €
    49.35 € / 60mL
    Save 2.70 €
    5% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium Phosphate Buffer is a reagent with very high buffering capacity that is widely used in molecular biology, biochemistry, and chromatography. It occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4).

  • Component of the FastDNA SPIN Kit for Soil and FastDNA SPIN Kit for Feces.
  • Sodium phosphate is a reagent with very high buffering capacity that is widely used in molecular biology, biochemistry, and chromatography.
  • It occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4).

Specifications

Content And Storage Store at Room Temperature (15°C to 30°C)
For Use With (Application) Sample Preparation, Sample Lysis, Nucleic Acid Isolation and Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
For Use With (Equipment) FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1
pH 7.8 to 8.2
Product Type Sodium Phosphate Buffer
Quantity 60 mL

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.