missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Sorbitol Dehydrogenase, From Sheep Liver
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| pH | Approximately 9.0 to 9.5 |
| Content And Storage | 0°C |
| Quantity | 5 mg |
| Product Type | Sorbitol Dehydrogenase, L-Iditol dehydrogenase, Polyol dehydrogenase, SDH |
| Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction