missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPIN Module and Recovery Tubes

Product Code. 12733517 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 Pk.
60 Pk.
Unit Size:
Pack of 100
Pack of 60
This item is not returnable. View return policy

Product Code. 12733517

Brand: MP Biomedicals™ 112080800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for optimal performance, providing the purest DNA faster than ever, and with no matrix carry-over. SPIN Module and Recovery Tubes are applied in molecular biology for their ability to separate particles from solutions in a few seconds.

  • Spin Module give faster and cleaner separations than traditional methods such as columns, filters, or simple centrifugation
  • When used with the GENECLEAN™ or MERmaid™ protocols, they combine the speed, efficiency, and selectivity of the GLASSMILK™ binding reaction with an even more rapid execution of the separation, washing, and elution steps
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Sample Type Lysate or other suspension
For Use With (Application) SPIN Module is applied in molecular biology for its ability to separate particles from solutions in a few seconds.
Product Type SPIN Module and Recovery Tubes
Quantity 100 Pk.
Sample Volume 500 to 700 μL liquid per centrifugation, Larger volumes require multiple loadings

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.