missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Stem Cell Factor, Human, Recombinant

Product Code. 11429921 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 μ g
2μg
Unit Size:
10µg
2µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429921 10 μ g 10µg
11419921 2μg 2µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429921 Supplier MP Biomedicals™ Supplier No. 0215869810
  • ONLINE EXCLUSIVE PRICE
    470.00 €
    445.20 € / 10µg
    Save 24.80 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stimulates the proliferation of bone marrow cells in 5-fluorouracil treated mice

  • Hematopoietic growth factor, also known as Mast Cell Growth Factor or c-kit ligand
  • Expressed in E. coli
  • ED50 is 20.0 ng/mL

Believed to stimulate the proliferation of myeloid, erythroid and lymphoid progenitors in bone marrow cultures and acts synergistically with colony stimulating factors, resulting in greater size and yields

Order Info

Supplied lyophilized

Specifications

Content And Storage -20°C
Product Type Stem Cell Factor
Formulation Sterile filtered then the protein was lyophilized with no additives
For Use With (Application) Believed to stimulate the proliferation of myeloid, erythroid and lymphoid progenitors in bone marrow cultures and acts synergistically with colony stimulating factors, resulting in greater size and yields
Purity or Quality Grade ≥98%
Quantity 10 μ g
Reactivity E. coli
Source Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.