missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Terrific Broth II, Dehydrated Media

Product Code. 11367699 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
454 g
1000 g
5000 g
Unit Size:
1kg
454g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367699 1000 g 1kg
11347699 454 g 454g
11387699 5000 g 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367699 Supplier MP Biomedicals™ Supplier No. 113046032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rich medium for growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Grade Molecular Biology
Product Type Microbiology Media
Formulation Yeast extract, Tryptone broth, Sodium Chloride and Vitamins
For Use With (Application) For growth of Escherichia coli
Quantity 1000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.