missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Wheat Germ Agglutinin, 5mg

Product Code. 11454186 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454186 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11454186 Supplier MP Biomedicals™ Supplier No. 08790162
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

isolated from wheat germ, dissolved in 0.05 M Tris-HCl, 0.15 M NaCl buffer, pH 8.6 and purified by affinity chromatography

Specifications

Quantity 5 mg
Form Powder
For Use With (Application) Immunoassay
Product Type Wheat germ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.