missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Species | Horse |
| Form | Lyophilized |
| Product Type | Horse Serum |
| Quantity | 2 mL |
| Sterilization Method | 0.2 μm filtration |
| Formulation | Delipidated, lyophilized from PBS with Azide |
| For Use With (Application) | Immunoelectrophoresis, ouchterlony immunodiffusion, and other immunoprecipitation techniques, blocking agent in fluorescence and enzyme immunoassays |
| pH | 7.3 |
| Protein Content | 63.5 mg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?