missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ YNB, Powdered Media: Without Ammonium Sulfate

Product Code. 11347809 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
227 g
454 g
1000 g
Unit Size:
1kg
227g
454g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347809 454 g 454g
11337809 227 g 227g
11357809 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347809 Supplier MP Biomedicals™ Supplier No. 114027022
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Can be prepared by adding a carbon source, the appropriate amino acids, and agar if necessary

  • Nitrogen-based yeast

Yeast cell culture, Gene expression, Transcription regulation

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Media Type YNB
Product Type Dehydrated Culture Media
Formulation Nitrogen Source: Ammonium Sulfate 5000mg/L; Vitamins: Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg; Trace Elements: Boric Acid 0.5mg, Copper Sulfate 0.04g, Potassium Iodide 0.1mg, Ferric Chloride 0.2mg, Manganese Sulfate 0.4mg, Sodium Molybdate 0.2mg, Zinc Sulfate 0.4mg; Salts: Magnesium Sulfate 500mg, Sodium Chloride 100mg, Potassium Phosphate monobasic 1000mg, Calcium Chloride 100mg
For Use With (Application) For vigorous growth of Saccharomyces cerevisiae
Quantity 454 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.