missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ YPD (YEPD) Agar, Powder

Product Code. 11337759 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1000 g
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337759 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11337759 Supplier MP Biomedicals™ Supplier No. 114001232
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rich medium for the growth of S. cereviseae in liquid medium

  • Contains dextrose, peptone, and yeast extract
  • Bulk powder packaging
Formulation
  • Contents/liter: 20g peptone, 10g yeast extract, 20g dextrose

Yeast Cell Culture

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Grade Molecular biology
Product Type Agar
Formulation Casein Peptone type-M 20g/L, Yeast extract 10g/L, Dextrose 20g/L
For Use With (Application) For growth of Saccharomyces cerevisiae in liquid medium
Quantity 1000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.