missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MPEP hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MPEP hydrochloride is a potent and highly selective non-competitive antagonist at the mGlu5 receptor subtype (IC50 = 36 nM) and a positive allosteric modulator at mGlu4 receptors. Centrally active following systemic administration in vivo. Reverses mechanical hyperalgesia in the inflamed rat hind paw.
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-6-(phenylethynyl)pyridine hydrochloride |
| CAS | 219911-35-0 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Target | Group I mGluR Antagonists |
| Molecular Formula | C14H11N.HCl |
| Formula Weight | Observed MW: 229.71 |
| Purity | 99% |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction