Description
- Time-resolved fluorescence immunoassay analyzer is a rapid, quantitative instrument that tests quantitative items such as cardiac markers, inflammation markers, hormones, vitamin D and diabetes.
- It is an automatic in-vitro diagnostic device that measures the concentration of analytes contained in blood, serum, and plasma samples.
- The immunofluorescence analyzer should be used only by medical professionals to carry out in-vitro diagnostic.
- It can be used in central laboratories, outpatient / emergency laboratories, clinical departments, and other medical service points such as community medical points and physical examination centers and can also be used in pre-hospital first aid ambulances.
- CE-marked
- Dedicated to time-resolved fluorescence immunoassay
- Compatible with different sample type: whole blood, serum and plasma
- Built-in printer
- Data storage: more than 10,000 test records
- Wide range of languages software
- For professional in-vitro diagnostic use
- The device allows you to test:
- Cardiac markers: CK-MB, Troponin I, Myoglobin and D-Dimer.
- Inflammation markers: C-Reactive Protein, Interleukin-6 and Procalcitonin.
- Diabetes and Vitamin D: HBA1c and 25-Hydroxyvitamin D.
- Hormones: B-HCG, LH, Progesterone, Prolactin, TSH, T3, T4, Free T3, Free T4.
Specifications
Specifications
| Type | Accuracy Reader |
| Sample Type | Blood |
| Memory | 10,000 test records |