missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MRPs-BCRP Vesicular Transport Assay Reagent Set

Product Code. 10121240 Shop All Gibco Products
Change view
Click to view available options
Quantity:
100 assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10121240 100 assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10121240 Supplier Gibco™ Supplier No. GM3010
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MRPs-BCRP Vesicular Transport Assay Reagent Set

The MRPs-BCRP Vesicular Transport Assay Reagent Set is a ready-to-use kit containing all reagents needed for vesicular transport assay except for the vesicles themselves. The reagent set can be used for vesicular transport assay using radioisotope-labeled or non-labeled compound. This kit has enough material to perform 100 vesicular transport assays.

Use The MRPs-BCRP Vesicular Transport Assay Reagent Set to:

  • Investigate the transporter interactions of your drug candidates
  • Assess potential for transporter-mediated drug-drug interactions
  • Obtain high quality results with a large signal to noise ratios

    Concept Behind ABC Transporter Vesicles
    ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate’s substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.

    Clear and Reliable Results
    Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these 'inside-out' vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.

    Kit Compatibility
    Our MRPs-BCRP Vesicular Transport Assay Reagent Set is designed for use with either MRP or BCRP Vesicles. The 100 vesicular transport assays that can be performed with this kit will require two MRP or BCRP Vesicle products.

    For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store at -20°C.
    Format Frozen
    Quantity 100 assays
    Shipping Condition Dry Ice
    Type Vesicular Transport Assay Reagent Set

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.