missing translation for 'onlineSavingsMsg'
Learn More

MRS 1334, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18065462
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18065462 50 mg 50mg
18025852 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065462 Supplier Tocris Bioscience Supplier No. 1385/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent, highly selective hA3 antagonist

Potent and highly selective antagonist for the human adenosine A3 receptor. Ki values are 2.69 nM at hA3, and > 100 μM at rat A1 and rat A2A receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192053-05-7
Molecular Formula C31H26N2O6
Molecular Weight (g/mol) 522.557
InChI Key WICWQHSRWZIPNB-AREMUKBSSA-N
Synonym 1, 4-Dihydro-2-methyl-6-phenyl-4-(phenylethynyl)-3, 5-pyridinedicarboxylic acid 3-ethyl-5-[(3-nitrophenyl)methyl] ester
PubChem CID 98072178
IUPAC Name 3-O-ethyl 5-O-[(3-nitrophenyl)methyl] (4R)-2-methyl-6-phenyl-4-(2-phenylethynyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C#CC2=CC=CC=C2)C(=O)OCC3=CC(=CC=C3)[N+](=O)[O-])C4=CC=CC=C4)C

Specifications

Quantity 50 mg
Formula Weight 522.56
Percent Purity >98%
Chemical Name or Material MRS 1334
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.