missing translation for 'onlineSavingsMsg'
Learn More

MRS-1706, MedChemExpress

Product Code. 16413808
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16413808

Brand: Medchem Express HY103186

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MRS-1706 is a potent and selective adenosine A2B receptor inverse agonist. MRS-1706 has Ki values of 1.39, 112, 157, and 230 nM for human A2B, A2A, A1 and A3 receptors respectively. MRS-1706 blocks adenosine-mediated cAMP induction.

MRS-1706 is a potent and selective adenosine A2B receptor inverse agonist. MRS-1706 has Ki values of 1.39, 112, 157, and 230 nM for human A2B, A2A, A1 and A3 receptors respectively. MRS-1706 blocks adenosine-mediated cAMP induction.

Specifications

Chemical Name or Material MRS-1706
CAS 264622-53-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H29N5O5
Quantity 10 mg
Solubility Information DMSO : 240 mg/mL (476.62 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C(C)=O)C=C1)COC2=CC=C(C3=NC(N(CCC)C(N(CCC)C4=O)=O)=C4N3)C=C2
Molecular Weight (g/mol) 503.55
Formula Weight 503.55
Percent Purity 98.23%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.