missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MRS 2211

Description
MRS 2211 is a competitive P2Y13 receptor antagonist (pIC50 = 5.97) that displays > 20-fold selectivity over P2Y1 and P2Y12 receptors.
Specifications
Specifications
| Chemical Name or Material | 2-[(2-Chloro-5-nitrophenyl)azo]-5-hydroxy-6-methyl-3-[(phosphonooxy)methyl]-4-pyridinecarboxaldehyde disodium salt |
| Assay Percent Range | >96% |
| Quantity | 50 mg |
| Solubility | Soluble to 70mM in water |
| Molecular Formula | C14H10N4O8Na2PCl |
| Formula Weight | Observed MW: 474.66 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?