missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MRTX 849

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MRTX 849 is a mutant-selective inhibitor of KRASG12C (IC50 = 142 nM). MRTX 849 covalently binds to KRASG12C at the cysteine at residue 12 and inhibits KRAS-dependent signal transduction by locking the protein in its inactive GDP-bound state. MRTX 849 induces pronounced tumor regression in 17 of 26 (65%) KRASG12C -positive cell lines and patient-derived xenograft models from multiple tumor types. It is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 4-[7-(8-Chloro-1-naphthalenyl)-5,6,7,8-tetrahydro-2-[[(2 S)-1-methyl-2-pyrrolidinyl]methoxy]pyrido[3,4-d]pyrimidin-4-yl]-1-(2-fluoro-1-oxo-2-propen-1-yl)-(2 S)-2-piperazineacetonitrile |
| CAS | 2326521-71-3 |
| Quantity | 10 mg |
| Target | Ras GTPase Inhibitors |
| Molecular Formula | C32H35ClFN7O2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction