missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MS 15203

Product Code. 18728078
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18728078

Brand: Tocris Bioscience™ 6299/25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and selective GPR171 partial agonist

MS 15203 is a potent and selective GPR171 partial agonist (EC50 = 90 nM; 28% inhibition of Forskolin activated adenylyl cyclase activity in rat hypothalamic membranes). Exhibits minimal binding to a panel of ∼70 GPCRs and 10 other receptors (<22% of radioligand displaced from the target receptors). Increases neuronal activity in the paraventricular nucleus. Increases food intake and body weight in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[(2-Methyl-1-oxo-2-propen-1-yl)amino]-1,3-benzenedicarboxylic acid
CAS 73912-52-4
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C12H11NO5
Formula Weight Observed MW: 249.22
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.