missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MS 15203
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
MS 15203 is a potent and selective GPR171 partial agonist (EC50 = 90 nM; 28% inhibition of Forskolin activated adenylyl cyclase activity in rat hypothalamic membranes). Exhibits minimal binding to a panel of ∼70 GPCRs and 10 other receptors (<22% of radioligand displaced from the target receptors). Increases neuronal activity in the paraventricular nucleus. Increases food intake and body weight in mice.
Specifications
Specifications
| Chemical Name or Material | 5-[(2-Methyl-1-oxo-2-propen-1-yl)amino]-1,3-benzenedicarboxylic acid |
| CAS | 73912-52-4 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C12H11NO5 |
| Formula Weight | Observed MW: 249.22 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction